missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DEAE-Dextran, chloride Form, MP Biomedicals™
$101.60 - $101.60
Chemical Identifiers
CAS | 9015-73-0 |
---|---|
MDL Number | MFCD00081561 |
Synonym | Diethylaminoethyl-dextrane |
IUPAC Name | Diethylaminoethyl dextran |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19513310
|
MP Biomedicals Inc
0219513310 |
10 g |
Each for $101.60
|
|
|||||
Chemical Identifiers
9015-73-0 | |
MFCD00081561 | |
Diethylaminoethyl-dextrane | |
Diethylaminoethyl dextran |
Specifications
Powder | |
MFCD00081561 | |
10 g | |
Diethylaminoethyl dextran | |
DEAE-Dextran |
9015-73-0 | |
White | |
Diethylaminoethyl-dextrane | |
500, 00 |