Decalcifying Solution B, Protocol, Fisher HealthCare™
  • Hydrochloric acid solution for rapid decalcification of bone
  • Compatible with hematoxylin and eosin staining procedures
Supplier:  Epredia 245683
 View more versions of this product
Catalog No. 23-245683
Explore available promotions
Description
Description
Specifications
Specifications
Fisher Scientific™ Protocol Decalcifying Solution B
Poly Bottle
4 L