Learn More
Decanoyl chloride, 98%
CAS: 112-13-0 | C10H19ClO | 190.711 g/mol
$71.66 - $231.26
Chemical Identifiers
| CAS | 112-13-0 |
|---|---|
| Molecular Formula | C10H19ClO |
| Molecular Weight (g/mol) | 190.711 |
| MDL Number | MFCD00000771 |
| InChI Key | IPIVAXLHTVNRBS-UHFFFAOYSA-N |
| Synonym | capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n |
| PubChem CID | 66982 |
| IUPAC Name | decanoyl chloride |
| SMILES | CCCCCCCCCC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1948622
|
Thermo Scientific Chemicals
A1948622 |
100 g |
N/A
|
|
|||||
|
AAA1948636
|
Thermo Scientific Chemicals
A1948636 |
500 g |
N/A
|
|
|||||
Description
Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
| 112-13-0 | |
| 190.711 | |
| IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
| 66982 | |
| CCCCCCCCCC(=O)Cl |
| C10H19ClO | |
| MFCD00000771 | |
| capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
| decanoyl chloride |
Specifications
| 112-13-0 | |
| 0.925 | |
| 106°C (223°F) | |
| C10H19ClO | |
| MFCD00000771 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| CCCCCCCCCC(=O)Cl | |
| 190.711 | |
| 190.71 | |
| Decanoyl chloride |
| -35°C | |
| 232°C | |
| Irritating | |
| 1.441 | |
| 100 g | |
| 507055 | |
| capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n | |
| IPIVAXLHTVNRBS-UHFFFAOYSA-N | |
| decanoyl chloride | |
| 66982 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 203-938-0
RTECSNumber : HE0875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only