Decon™ AquaPur™ Purified USP-Grade Water
For use in laboratory or production applications
$94.37 - $1968.87
Alert:
Not to be used for parenteral administration. Not for use in sterile irrigation, injection, infusion, or as a potable product.For manufacturing and laboratory use only.
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
04-355-126
|
Decon™
9355 |
55 gal. |
Each for $1,968.87
|
|
|||||
04-355-124
|
Decon™
DC9301 |
1 gal. |
Each for $94.37
Case of 4 Each for $377.48
|
|
|||||
04-355-125
|
Decon™
9305 |
5 gal. |
Each for $449.63
|
|
|||||
Description
Tested to meet USP specifications and packaged in convenient point-of-use packaging. Proprietary manufacturing process ensures AquaPur Purified Water maintains microbial specification through expiration date.
Meets these specifications:
- USP Water Conductivity, USP 645 (at manufacture)
- USP Total Organic Carbon, USP 643 < 0.5ppm
- Oxidizable Substances: Conforms (at manufacture)
- Total Microbial Count: < 100cfu/mL