Decon™ ELIMINase™ Decontaminant
Eliminates RNase, DNase, and DNA from laboratory surfaces
Supplier: Decon™ DC1102
Description
- Safe on glass, plastics, stainless steel, pipetters, gel boxes, and benchtops
- Also acts as a radioactive decontaminant
- Leaves no residue that could degrade RNA and DNA samples or interfere with subsequent enzymatic reaction
- Works instantly
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Specifications
ELIMINase™ Laboratory Decontaminant | |
32 oz. | |
950 mL | |
Eliminates RNase, DNase & DNA contamination |