missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decon™ Foot Operated Alcohol Dispenser
Easily dispenses Cidehol while keeping hands free
Supplier: Decon™ 8310
Description
- Can hold a 473.2mL (16 oz.) bottle
- For CiDehol ST 70%, CiDehol 70, CiDehol ST 99%, SaniHol ST, and SaniHol 70 sterile alcohols
Specifications
CiDehol™ 70 Alcohol Dispenser | |
Used to clean surfaces and hoods in microbiology labs and tissue culture labs | |
120°F |
70% Isopropyl Alcohol, 30% purified water | |
16 oz. Bottles of CiDehol™ ST 70%, CiDehol™ 70, CiDehol™ ST 99%, SaniHol ST, and SaniHol 70 sterile Alcohols | |
48.8°C |