Decon™ LeLoop™ Inoculating Loops
More economical than platinum or disposable plastic loops
Supplier:  Decon™ MP19725
 View more versions of this product
Catalog No. 22-032090
Explore available promotions
Description
Description
More economical than platinum or disposable plastic loops
Reusable loops are made of high-quality twisted nichrome wire.
  • Loops slide smoothly across agar surfaces without tearing or gouging
  • The nichrome wire cools rapidly, saving time
  • Loops are available calibrated or noncalibrated
  • Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
  • LeLoop™ loops come without holders, which are available separately
Specifications
Specifications
0.08 in.
Nichrome
Inoculating Wire Loop
0.22 cm
2.45 μL
No
Videos
SDS
Documents
Documents
Accessories