missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decon™ LeStreak™ Microstreakers
Perfect for inoculating or transferring microbiological samples
Supplier: Decon™ MP1845
Description
Perfect for inoculating or transferring microbiological samples
- Use to inoculate or transfer microbiological samples
- Made of high-quality twisted nichrome wire permanently attached to lightweight insulated aluminum handles
- Slide smoothly across agar surfaces without tearing or gouging
- Nichrome wire cools rapidly after flaming
- 1 and 10μL loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test
- Noncalibrated microstreakers are color coded for easy identification
Certifications
Calibration certificates provided for 1μL and 10μL loops
Specifications
0.08 in. | |
Nichrome | |
Blue | |
Nichrome wire loop permanently attached to blue insulated aluminum holder |
0.22 cm | |
2.45 μL | |
Inoculating Streaker | |
No |