Deep-Well Projection Slides
View specimens by either microscope or projector.
Supplier:  Customer Channels Group (Ccg) S17889
Catalog No. S17889
Explore available promotions
Description
Description
Slides hold live aquatic specimens in the well and permit students to view them under a microscope or by means of a 35mm or overhead projector. Chemistry students can view crystal formation.
Specifications
Specifications
Microscope Slide
Viewing under microscope or 35mm overhead projector with cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'