OHAUS™ Defender 5000 Bench Scales, 12 x 12 Inch Platform
Defender 5000 Multifunctional Bench Scale features an indicator that guides the user with on-screen prompts and soft keys guides
Supplier:  OHAUS™ 30461648
 View more versions of this product
Catalog No. 01922231
Explore available promotions
Description
Description
  • Results shown clearly on the large backlit display
  • Alpha-numeric keypad and function keys allow for a simplified user experience
Specifications
Specifications
CE/CSA
12.5 kg, 25 lb.
0.5 g
Stainless Steel Pan and Powder Coated Steel Frame Base
14°F to 104°F
12 x 12 in.
45 cm
9 kg
Standard RS232, MicroSD Card (included). Optional Ethernet, WiFi/Bluetooth, 2nd RS232/RS485/USB, Analog Output, 2 In/4 Out Digital I/O available.
Front Mounted ABS Plastic LCD Indicator
AC Adapter
2 sec.
Legal For Trade
48 cm
36 cm
Yes – Class III