missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dehydroacetic acid, 98%, Thermo Scientific Chemicals

$47.23 - $367.14

Chemical Identifiers

CAS 520-45-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00066709
InChI Key PGRHXDWITVMQBC-UHFFFAOYSA-N
Synonym dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
PubChem CID 122903
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
SMILES CC1=CC(=O)C(C(=O)O1)C(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1782718
View Documents
Thermo Scientific Chemicals
A1782718
50 g N/A
 
AAA1782730
View Documents
Thermo Scientific Chemicals
A1782730
250 g N/A
 
AAA178270B
View Documents
Thermo Scientific Chemicals
A178270B
1000 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

520-45-6
168.148
PGRHXDWITVMQBC-UHFFFAOYSA-N
122903
CC1=CC(=O)C(C(=O)O1)C(=O)C
C8H8O4
MFCD00066709
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
3-acetyl-6-methylpyran-2,4-dione

Specifications

520-45-6
269°C to 270°C
C8H8O4
50 g
14,2865
PGRHXDWITVMQBC-UHFFFAOYSA-N
3-acetyl-6-methylpyran-2,4-dione
122903
98%
109°C to 113°C
157°C (314°F)
MFCD00066709
6129
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
CC1=CC(=O)C(C(=O)O1)C(=O)C
168.148
168.15
Dehydroacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 208-293-9

RTECSNumber : UP8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only