Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Supplier:  Thermo Scientific™ D0803
 View more versions of this product
Catalog No. 09-034
Explore available promotions
Description
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
Specifications
Harvey DI+ Replacement Cartridge
Two Bed Ion Exchange
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
1760 grains
Cartridge for high capacity; two-bed ion exchange; full size
Deionizer Cartridge