Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Supplier:  Thermo Scientific™ D0836
 View more versions of this product
Catalog No. 09-034-5C
Explore available promotions
Description
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
Specifications
Cartridge for pretreatment macropure; macroreticular, activated carbon; full size
Macroreticular, Activated Carbon
B-Pure™
2000 gal.
Pretreatment Macropure
Deionizer Cartridge