missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Demineralizer Cartridges

Special resin dye changes color when cartridge is exhausted.

$242.59 - $242.59

Specifications

For Use With (Application) Water purification
View More Specs

Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
S508171
View Documents
Thermo Scientific™
D8900
Cartridge Holder
Each for $242.59
 
Description

Description

Hose nipples on each end of these cartridges easily connect to your water supply with ordinary lab tubing (0.3 in. I.D. [9.5mm]). Resins in the deionization cartridge contain a special dye that changes color when the cartridge is exhausted.

High-Capacity Cartridge

  • Has a two-bed demineralizer with selected cation and anion resin beds
  • Delivers more liters per cartridge than mixed-bed types
  • Average resistance of effluent is 175kilohm·cm
  • Total ion-exchange capacity is 1725 grains (as NaCl)
UltraPure Cartridge
  • Produces high-resistance demineralized water from tap water
  • Increases purity of water from central demineralizer
  • Total ion-exchange capacity is 1200 grains (as NaCl), 849 grains (as NaCl) to 1 megohm·cm endpoint
  • pH is usually between 6.8 and 7.2
  • Water is largely free of silica and CO2

Specifications

Specifications

Water purification
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'