missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Denatonium benzoate, Thermo Scientific Chemicals

The bitterest compound known

$62.63 - $759.68

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Synonym unii-m5ba6gaf1o
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6104803
View Documents
Thermo Scientific Chemicals
J6104803
1 g
Each for $62.63
 
AAJ6104806
View Documents
Thermo Scientific Chemicals
J6104806
5 g
Each for $186.27
 
AAJ6104814
View Documents
Thermo Scientific Chemicals
J6104814
25 g
Each for $759.68
 
Description

Description

The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
Specifications

Chemical Identifiers

3734-33-6
C28H34N2O3
unii-m5ba6gaf1o

Specifications

3734-33-6
1 g,1 g
164°C to 169°C
446.59
White
unii-m5ba6gaf1o
Soluble in isopropyl alcohol,methanol. Slightly soluble in acetone. Insoluble in ether.
C28H34N2O3
Solid
Denatonium benzoate
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

EINECSNumber : 223-095-2

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only