missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethanol, 70% Solution, Molecular Biology Grade, Denatured, Fisher BioReagents™
Used as a cleaning agent for labortatory use
Supplier: Thermo Fisher Scientific BP82031GAL
Description
- DNase, and Protease tested to verify absence of hydrolytic enzymes
- Combine this blend with commercial “mini-prep“ kits or traditional extraction
- ACS grade Absolute Ethanol used in preparation of solution
Specifications
70% Denatured Ethanol Solution (Molecular Biology Grade) | |
64-17-5,67-63-0,7732-18-5 | |
78.4°C | |
Poly Bottle | |
≤ 0.001% | |
44.6 mm Hg at 20°C (68°F) | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 |
-144°C | |
Colorless | |
1 gal. | |
Liquid | |
C2H6O | |
3%-7% (GC-TCD) | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
Molecular Biology |
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |