missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage
A convenient, streamlined process to maximize sample storage. Our freezer racking and tubing solutions are designed to hold tubes ranging from 200μL to 5.0μL.
Supplier: Thermo Scientific™ 1950842
Description
A convenient, streamlined process to maximize sample storage in your laboratory. Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200μL up to 5.0μL, offering maximized sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.
- Warranty varies by country, please contact us for details
Specifications
Tube Rack (Cryogenic and Ultra-Low Chest Freezers) | |
Silver | |
Rectangular | |
Thermo Scientific CryoExtra 94 (holds 92 racks) | |
5.3 in. | |
33 in. | |
9.3 cm |
Racks for Tubes (cryogenic and ultra-low chest freezers) | |
Plates (22/Rack) | |
Thermo Scientific Matrix 200µL Screwtop universal tubes, 500µL storage tubes, 500µL storage tubes with DuraSeal, 0.5mL Nunc Cryobank tubes | |
13.5 cm | |
83.8 cm | |
3.6 in. |