missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Deoxyribonuclease I, MP Biomedicals™

$413.60 - $413.60

Chemical Identifiers

CAS 9003-98-9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10057510
View Documents
MP Biomedicals Inc
0210057510
10mg
Each for $413.60
 
Description

Description

Specifications

Chemical Identifiers

9003-98-9

Specifications

White
MFCD00130918
Deoxyribonuclease
10mg
Powder
9003-98-9
*
0.00
Deoxyribonuclease I
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'