DWK Life Sciences Kimble™ Desiccator with Plastic Top Knob 250x330
Excellent for storage of hygroscopic chemicals, and to prevent adsorption of moisture when using crucibles, weighing bottles, and watch glasses.
Supplier:  DWK Life Sciences 31050250
Catalog No. 12-141-432
Explore available promotions
Description
Description
Excellent for storage of hygroscopic chemicals, and to prevent adsorption of moisture when using crucibles, weighing bottles, and watch glasses.
  • Plastic screw-thread top cap for easy cover removal
  • Recessed base for containment of desiccant
  • Includes metal plate
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II, requirements
Specifications
Specifications
Desiccator with Plastic Top Knob