missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Desloratadine 98.0+%, TCI America™

$67.14 - $395.00

Chemical Identifiers

CAS 100643-71-8
Molecular Formula C19H19ClN2
Molecular Weight (g/mol) 310.825
MDL Number MFCD00871949
InChI Key JAUOIFJMECXRGI-UHFFFAOYSA-N
PubChem CID 124087
ChEBI CHEBI:291342
IUPAC Name 8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES C1CC2=C(C=CC(=C2)Cl)C(=C3CCNCC3)C4=C1C=CC=N4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D3787100MG
View Documents
TCI America
D3787100MG
100 mg N/A
 
D37871G
View Documents
TCI America
D37871G
1 g N/A
 
Description

Description

Specifications

Chemical Identifiers

100643-71-8
310.825
JAUOIFJMECXRGI-UHFFFAOYSA-N
CHEBI:291342
C1CC2=C(C=CC(=C2)Cl)C(=C3CCNCC3)C4=C1C=CC=N4
C19H19ClN2
MFCD00871949
124087
8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine

Specifications

100643-71-8
C19H19ClN2
100 mg
C1CC2=C(C=CC(=C2)Cl)C(=C3CCNCC3)C4=C1C=CC=N4
310.825
CHEBI:291342
≥98.0% (HPLC,T)
Desloratadine
157°C
MFCD00871949
JAUOIFJMECXRGI-UHFFFAOYSA-N
8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
124087
310.83
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DE8011000

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'