missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DETA NONOate, 95%, Thermo Scientific Chemicals

$155.88 - $385.65

Chemical Identifiers

CAS 146724-94-9
Molecular Formula C4H13N5O2
Molecular Weight (g/mol) 163.2
MDL Number MFCD00278808
InChI Key HMRRJTFDJAVRMR-UHFFFAOYSA-N
Synonym deta nonoate, noc-18, deta-nonoate, diethylenetriamine nonoate, 2,2'-hydroxynitrosohydrazino bis-ethanamine, aeed/no, 1,1-bis 2-aminoethyl-2-hydroxy-3-oxotriazane, 3,3-bis aminoethyl-1-hydroxy-2-oxo-1-triazene, detanonoate, 1-hydroxy-2-oxo-3,3-bis 3-aminoethyl-1-triazene
PubChem CID 1606
ChEBI CHEBI:50154
IUPAC Name N-[bis(2-aminoethyl)amino]-N-hydroxynitrous amide
SMILES C(CN(CCN)N(N=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328650250
SDS View Documents
Thermo Scientific Chemicals
328650250
25 mg Glass bottle
Each for $155.88
 
AC328651000
SDS View Documents
Thermo Scientific Chemicals
328651000
100 mg Glass bottle
Each for $385.65
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

146724-94-9
163.2
HMRRJTFDJAVRMR-UHFFFAOYSA-N
1606
N-[bis(2-aminoethyl)amino]-N-hydroxynitrous amide
C4H13N5O2
MFCD00278808
deta nonoate, noc-18, deta-nonoate, diethylenetriamine nonoate, 2,2'-hydroxynitrosohydrazino bis-ethanamine, aeed/no, 1,1-bis 2-aminoethyl-2-hydroxy-3-oxotriazane, 3,3-bis aminoethyl-1-hydroxy-2-oxo-1-triazene, detanonoate, 1-hydroxy-2-oxo-3,3-bis 3-aminoethyl-1-triazene
CHEBI:50154
C(CN(CCN)N(N=O)O)N

Specifications

146724-94-9
White
95%
C4H13N5O2
25 mg
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol
C(CN(CCN)N(N=O)O)N
163.2
CHEBI:50154
95%
DETA NONOate
98.0°C to 106.0°C
Authentic
Glass bottle
MFCD00278808
deta nonoate, noc-18, deta-nonoate, diethylenetriamine nonoate, 2,2'-hydroxynitrosohydrazino bis-ethanamine, aeed/no, 1,1-bis 2-aminoethyl-2-hydroxy-3-oxotriazane, 3,3-bis aminoethyl-1-hydroxy-2-oxo-1-triazene, detanonoate, 1-hydroxy-2-oxo-3,3-bis 3-aminoethyl-1-triazene
HMRRJTFDJAVRMR-UHFFFAOYSA-N
N-[bis(2-aminoethyl)amino]-N-hydroxynitrous amide
1606
163.2
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only