Learn More
Dexamethasone 21-phosphate disodium salt, 98%, Thermo Scientific Chemicals
Dexamethasone 21-phosphate disodium salt, CAS # 2392-39-4, is a synthetic adrenal glucocorticoid with potent anti-inflammatory activity.
$243.56 - $746.55
Chemical Identifiers
CAS | 2392-39-4 |
---|---|
Molecular Formula | C22H28FNa2O8P |
Molecular Weight (g/mol) | 516.41 |
MDL Number | MFCD00079105 |
InChI Key | PLCQGRYPOISRTQ-FCJDYXGNSA-L |
Synonym | dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron |
PubChem CID | 16961 |
ChEBI | CHEBI:4462 |
IUPAC Name | disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
SMILES | CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6408303
|
Thermo Scientific Chemicals
J6408303 |
1 g |
Each for $243.56
|
|
|||||
AAJ6408306
|
Thermo Scientific Chemicals
J6408306 |
5 g |
Each for $746.55
|
|
|||||
Description
Dexamethasone 21-phosphate disodium salt acts as an inducer of apoptosis and inhibitor of the sodium phosphate symporter. It is used in the treatment of emetic effects of cancer therapy and epicondylitis. Further, it serves as an activator of food, medicine, glucocorticoid and anti inflammatory.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2392-39-4 | |
516.41 | |
PLCQGRYPOISRTQ-FCJDYXGNSA-L | |
16961 | |
disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
C22H28FNa2O8P | |
MFCD00079105 | |
dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron | |
CHEBI:4462 | |
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] |
Specifications
2392-39-4 | |
White | |
MFCD00079105 | |
6473066 | |
dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron | |
PLCQGRYPOISRTQ-FCJDYXGNSA-L | |
disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate | |
16961 | |
516.4 | |
Powder |
233°C to 240°C | |
C22H28FNa2O8P | |
1 g | |
142943 | |
Soluble in water. Slightly soluble in ethanol. Insoluble in dichloromethane. | |
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] | |
516.41 | |
CHEBI:4462 | |
98% | |
Dexamethasone 21-phosphate disodium salt |
Safety and Handling
Harmful if swallowed, Suspected of damaging the unborn child, May cause damage to organs through prolonged or repeated exposure
Acute toxicity (category 4), Reproductive toxicity (category 2), Specific target organ toxicity after repeated exposure (category 2)
EINECSNumber : 219-243-0
RTECSNumber : TU4056000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only