missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dexamethasone acetate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6369106

 View more versions of this product

Catalog No. AAJ6369106


Explore available promotions

Description

Description

Dexamethasone has many uses in the treatment of cancer. Dexamethasone relieves inflammation in various parts of the body. To treat or prevent allergic reactions. As treatment of certain kinds of autoimmune diseases, skin conditions, asthma and other lung conditions. As treatment for a variety of cancers, such as leukemia, lymphoma, and multiple myeloma. It is also used for replacement therapy in adrenal insufficiency and in a screening test for the diagnosis of cushing's syndrome. Dexamethasone Acetate has also been determined to markedly decrease the number of activated macrophages. In addition, Dexamethasone Acetate has been observed to significantly decrease mRNA levels of TPH2 (tryptophan hydroxylase-2) in the raphe nuclei of C57/Bl6 mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble at 100 mg/ml in acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Specifications

Dexamethasone acetate
240°C
C24H31FO6
5 g
142943
Soluble at 100 mg/ml in acetone
C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O
434.50
434.5
1177-87-3
White
MFCD00027407
2342608
dexamethasone acetate, 11b,16a-9-fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione 21-acetate
AKUJBENLRBOFTD-RPRRAYFGSA-N
2-[(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
40469526
Powder

Chemical Identifiers

1177-87-3
434.50
AKUJBENLRBOFTD-RPRRAYFGSA-N
40469526
C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O
C24H31FO6
MFCD00027407
dexamethasone acetate, 11b,16a-9-fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione 21-acetate
2-[(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Videos
Safety and Handling

Safety and Handling

Suspected of damaging the unborn child, May cause an allergic skin reaction, May cause damage to organs through prolonged or repeated exposure

Reproductive toxicity (category 2), Skin sensitisation (category 1), Specific target organ toxicity after repeated exposure (category 2)

EINECSNumber : 214-646-8

RTECSNumber : TU4050000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only