missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dextran Sulfate Sodium Salt (White to Off-white Powder), Fisher BioReagents
Supplier: Fisher BioReagents BP1585500
Description
Dextran Sulfate accelerates the rate of hybridization of DNA or RNA bound to membranes and may be used in Southern and Northern blotting procedures.Specifications
White | |
Solid | |
(C6H7O5)m(C6H7O5)n | |
15, 2953 | |
500 g | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
40,000 | |
CHEBI:116735 | |
7.5% max. |
Dextran Sulfate Sodium Salt | |
9011-18-1 | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
ethyl 4-aminobenzoate | |
2337 | |
Pass Test | |
Poly Bottle |
Chemical Identifiers
9011-18-1 | |
40,000 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
2337 | |
ethyl 4-aminobenzoate |
(C6H7O5)m(C6H7O5)n | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
CHEBI:116735 | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT