missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextrin, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 406281000

 View more versions of this product

Catalog No. AC406281000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White to Yellow
Crystalline Powder
C18H32O16
MFCD00081554
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
100 g
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
504.438
162.07
Plastic bottle
Dextrin
9004-53-9
(C6H10O5)x
15, 2955
Solubility in water: soluble in hot water. Other solubilities: insoluble in ether and alcohols
FYGDTMLNYKFZSV-MRCIVHHJSA-N
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
62698
Authentic

Chemical Identifiers

9004-53-9
504.438
FYGDTMLNYKFZSV-MRCIVHHJSA-N
62698
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
C18H32O16
MFCD00081554
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Safety and Handling

Safety and Handling

EINECSNumber : 232-675-4

RTECSNumber : HH9450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only