missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextromethorphan hydrobromide, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6173206

 View more versions of this product

Catalog No. AAJ6173206

Explore available promotions

Description

Description

It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

Solubility
Soluble in water (40 mM), and alcohol (1:10).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Dextromethorphan hydrobromide
125°C to 127°C
C18H26BrNO
5 g
14,8091
Soluble in water (40 mM),and alcohol (1:10).
CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br
5464025
Solid
125-69-9
White
MFCD02173901
6453793
dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
MISZALMBODQYFT-URVXVIKDSA-N
352.316
352.32

Chemical Identifiers

125-69-9
352.316
MISZALMBODQYFT-URVXVIKDSA-N
5464025
C18H26BrNO
MFCD02173901
dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 204-750-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only