missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dextrose, Monohydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 191005
Specifications
Dextrose | |
1496309 | |
C18H30N2O14 | |
2.5 kg | |
nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate | |
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1 | |
498.44 | |
BAKER ANALYZED™ Reagent |
Monohydrate | |
Poly Bottle | |
MFCD00150899 | |
1L = 1.54kg | |
LDMPZNTVIGIREC-HYRPJEHINA-N | |
(2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate | |
198.17 | |
Powder |
Chemical Identifiers
1496309 | |
MFCD00150899 |