missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dextrose, Monohydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 191001
Specifications
Dextrose | |
1496309 | |
C6H14O7 | |
1L = 1.54kg | |
OSNSWKAZFASRNG-WNFIKIDCSA-N | |
(2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate | |
133126654 | |
BAKER ANALYZED™ Reagent |
Monohydrate | |
Poly Bottle | |
500 g | |
6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p. | |
C(C1C(C(C(C(O1)O)O)O)O)O.O | |
198.17 | |
198.17 | |
Powder |
Chemical Identifiers
1496309 | |
133126654 |