Thermo Scientific™ DH5α Competent Cells
Chemically competent E. coli cells recommended for routine subcloning into plasmid vectors. Subcloning efficiency cells are not suitable for the generation of cDNA libraries.
Supplier: Thermo Scientific™ EC0111
Description
- The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gall
- Transformation efficiency: >1x106 cfu/μg pUC19 DNA
- Suitable for routine cloning applicationsConvenient economical packaging
- Contain genetic markers that allow for blue/white colony screening
- DH5α Competent Cells for Subcloning are suitable for the standard DNA cloning applications:
- DNA cloning following PCR or restriction digestion reactions
- Subcloning (transferring DNA from a parent vector to a destination vector)
- Plasmid isolation
- Genotype: F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1
- For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemically Competent | |
Dry Ice (-80°C) | |
E. coli | |
Cloning |
Lacks F' Episome | |
DH5α | |
Tube | |
20 Reactions |