missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-2-pyridyl thionocarbonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 390880010

 View more versions of this product

Catalog No. AC390880010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-2-pyridyl thionocarbonate
96°C to 99°C
Authentic
Glass bottle
MFCD00074870
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2
232.26
232.26
Crystalline Powder
96989-50-3
Yellow to Brown
97%
C11H8N2O2S
1 g
IKYOVSVBLHGFMA-UHFFFAOYSA-N
dipyridin-2-yloxymethanethione
719784
97%

Chemical Identifiers

96989-50-3
232.26
IKYOVSVBLHGFMA-UHFFFAOYSA-N
719784
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2
C11H8N2O2S
MFCD00074870
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate
dipyridin-2-yloxymethanethione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only