Learn More
Di-n-butyl ether, 99+%, extra pure
CAS: 142-96-1 | C8H18O | 130.23 g/mol
$180.02 - $516.28
Chemical Identifiers
| CAS | 142-96-1 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00009461 |
| InChI Key | DURPTKYDGMDSBL-UHFFFAOYSA-N |
| Synonym | di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether |
| PubChem CID | 8909 |
| IUPAC Name | 1-butoxybutane |
| SMILES | CCCCOCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC149690010
|
Thermo Scientific Chemicals
149690010 |
1 L | Glass Bottle |
N/A
|
|
||||
|
AC149690025
|
Thermo Scientific Chemicals
149690025 |
2.5 L | Glass Bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 142-96-1 | |
| 130.23 | |
| DURPTKYDGMDSBL-UHFFFAOYSA-N | |
| 8909 | |
| CCCCOCCCC |
| C8H18O | |
| MFCD00009461 | |
| di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
| 1-butoxybutane |
Specifications
| 142-96-1 | |
| 0.7680g/mL | |
| 25°C | |
| 99% min. (GC) | |
| C8H18O | |
| [CH3(CH2)3]2O | |
| 1 L | |
| 0.768 | |
| di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
| DURPTKYDGMDSBL-UHFFFAOYSA-N | |
| 1-butoxybutane | |
| 8909 | |
| 130.23 | |
| Extra Pure | |
| Di-n-butyl ether |
| -95.0°C | |
| 141.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.3978 to 1.3998 | |
| MFCD00009461 | |
| 01, 369 | |
| 15, 1572 | |
| Solubility in water: 0.03% (20°C). Other solubilities: miscible with ethanol,ether,acetone | |
| CCCCOCCCC | |
| 130.23 | |
| 0.69 mPa.s (20°C) | |
| 99+% | |
| Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN
GHS Signal Word: Warning
EINECSNumber : 205-575-3
RUO – Research Use Only