Learn More
Di-n-butyl ether, 99%, Thermo Scientific Chemicals
$57.15 - $176.23
Chemical Identifiers
CAS | 142-96-1 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.23 |
MDL Number | MFCD00009461 |
InChI Key | DURPTKYDGMDSBL-UHFFFAOYSA-N |
Synonym | di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether |
PubChem CID | 8909 |
IUPAC Name | 1-butoxybutane |
SMILES | CCCCOCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA16305AP
|
Thermo Scientific Chemicals
A16305AP |
500 mL |
Each for $57.15
|
|
||||||
AAA163050F
|
Thermo Scientific Chemicals
A163050F |
2500 mL |
Each for $176.23
|
|
||||||
Description
Di-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-butyl ether is used as a solvent for Grignard, Witting and alkyl lithium reactions. It is also used as a solvent for oils and fats and some natural and synthetic resins. It is considered as a replacement for terathydofuran in organic synthesis due to its less water and peroxide and high boiling point. In the pharmaceutical industry, it is used in the manufacturing process of active pharmaceutical ingredient such as procarbazine and cefaclor. In addition to this, it is used as an important solvent for the application of the coating.
Solubility
Miscible with alcohol, ether and acetone. Immiscible with water.
Notes
Air and light sensitive. Incompatible with oxidizing agents and strong acids.
Chemical Identifiers
142-96-1 | |
130.23 | |
DURPTKYDGMDSBL-UHFFFAOYSA-N | |
8909 | |
CCCCOCCCC |
C8H18O | |
MFCD00009461 | |
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
1-butoxybutane |
Specifications
-95°C | |
0.764 | |
500 mL | |
1.399 | |
MFCD00009461 | |
1732752 | |
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether | |
DURPTKYDGMDSBL-UHFFFAOYSA-N | |
1-butoxybutane | |
8909 | |
99% |
142-96-1 | |
141°C to 143°C | |
25°C (77°F) | |
C8H18O | |
UN1149 | |
14,1569 | |
Miscible with alcohol,ether and acetone. Immiscible with water. | |
CCCCOCCCC | |
130.23 | |
130.23 | |
Di-n-butyl ether |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIBUTYL ETHERS
EINECSNumber : 205-575-3
RTECSNumber : EK5425000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only