Learn More
Di-n-butyl phthalate, 99%, Thermo Scientific Chemicals
$43.53 - $339.27
Chemical Identifiers
CAS | 84-74-2 |
---|---|
Molecular Formula | C16H22O4 |
Molecular Weight (g/mol) | 278.35 |
MDL Number | MFCD00009441 |
InChI Key | DOIRQSBPFJWKBE-UHFFFAOYSA-N |
Synonym | dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db |
PubChem CID | 3026 |
ChEBI | CHEBI:34687 |
IUPAC Name | 1,2-dibutyl benzene-1,2-dicarboxylate |
SMILES | CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1325736
|
Thermo Scientific Chemicals
A1325736 |
500 g |
Each for $43.53
|
|
|||||
AAA132570E
|
Thermo Scientific Chemicals
A132570E |
2500 g |
Each for $114.01
|
|
|||||
AAA132570C
|
Thermo Scientific Chemicals
A132570C |
10,000 g |
Each for $339.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a general purpose monomeric phthalate with excellent solubilizing characteristics. It is commonly used as an additive to adhesives and printing inks. It also finds utility in shower curtains, raincoats and vinyl coated fabrics.
Solubility
Soluble in water. (0.4 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
84-74-2 | |
278.35 | |
DOIRQSBPFJWKBE-UHFFFAOYSA-N | |
3026 | |
1,2-dibutyl benzene-1,2-dicarboxylate |
C16H22O4 | |
MFCD00009441 | |
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db | |
CHEBI:34687 | |
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC |
Specifications
84-74-2 | |
1.045 | |
171°C (340°F) | |
C16H22O4 | |
MFCD00009441 | |
UN3082 | |
14,3035 | |
Soluble in water. (0.4g/L) | |
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC | |
278.35 | |
CHEBI:34687 | |
99% |
-35°C | |
339°C to 340°C | |
Odorless | |
1.492 | |
500 g | |
1914064 | |
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db | |
DOIRQSBPFJWKBE-UHFFFAOYSA-N | |
1,2-dibutyl benzene-1,2-dicarboxylate | |
3026 | |
278.35 | |
Di-n-butyl phthalate |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
H336-H360Df-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 201-557-4
RTECSNumber : TI0875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only