Learn More
Di-n-butylamine, 98+%, Thermo Scientific Chemicals
$74.51 - $266.62
Chemical Identifiers
CAS | 111-92-2 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD00009429 |
InChI Key | JQVDAXLFBXTEQA-UHFFFAOYSA-N |
Synonym | dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine |
PubChem CID | 8148 |
IUPAC Name | N-butylbutan-1-amine |
SMILES | CCCCNCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA11671AP
|
Thermo Scientific Chemicals
A11671AP |
500 mL |
Each for $74.51
|
|
||||||
AAA116710F
|
Thermo Scientific Chemicals
A116710F |
2500 mL |
Each for $266.62
|
|
||||||
Description
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
Solubility
Miscible with water, ether, ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.
Chemical Identifiers
111-92-2 | |
129.247 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
8148 | |
CCCCNCCCC |
C8H19N | |
MFCD00009429 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
N-butylbutan-1-amine |
Specifications
111-92-2 | |
0.767 | |
39°C (102°F) | |
C8H19N | |
[CH3(CH2)3]2NH | |
500 mL | |
506001 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
N-butylbutan-1-amine | |
8148 | |
≥98% | |
Di-n-butylamine |
°C to 62°C | |
159°C to 160°C | |
≥98% | |
1.4169 | |
MFCD00009429 | |
UN2248 | |
14,3032 | |
Miscible with water,ether,ethanol and acetone. | |
CCCCNCCCC | |
129.247 | |
129.25 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H311-H330-H314-H318-H226
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H311-H314-H330
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DI-N-BUTYLAMINE
EINECSNumber : 203-921-8
RTECSNumber : HR7780000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only