Learn More
Di-n-butylamine, 99%, Thermo Scientific Chemicals
Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL
Supplier: Thermo Scientific Chemicals 112950010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-n-butylamine | |
-62.0°C | |
159.0°C | |
Authentic | |
Glass Bottle | |
1.4160 to 1.4180 | |
MFCD00009429 | |
04, 157 | |
0.76 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
N-butylbutan-1-amine | |
8148 | |
99% |
111-92-2 | |
0.7600g/mL | |
39°C | |
99% | |
C8H19N | |
[CH3(CH2)3]2NH | |
1 L | |
01,209 | |
15, 3037 | |
Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons | |
CCCCNCCCC | |
129.247 | |
129.24 | |
Liquid |
Chemical Identifiers
111-92-2 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
8148 | |
CCCCNCCCC |
MFCD00009429 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
N-butylbutan-1-amine |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only