Learn More
Di-n-butylamine, 99%, Thermo Scientific Chemicals
Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL
Supplier: Thermo Scientific Chemicals 112950025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-n-butylamine | |
-62.0°C | |
159.0°C | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00009429 | |
04, 157 | |
0.76 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
N-butylbutan-1-amine | |
8148 | |
129.24 | |
Liquid |
111-92-2 | |
0.7600g/mL | |
39°C | |
98.5% min. (GC) | |
C8H19N | |
[CH3(CH2)3]2NH | |
2.5 L | |
01,209 | |
15, 3037 | |
Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons | |
CCCCNCCCC | |
129.24 | |
0.9 mPa.s (20°C) | |
99% |
Chemical Identifiers
111-92-2 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
8148 | |
CCCCNCCCC |
MFCD00009429 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
N-butylbutan-1-amine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre
GHS Signal Word: Danger
EINECSNumber : 203-921-8
RUO – Research Use Only