missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-n-butylamine, 99%, Thermo Scientific Chemicals

Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL

Supplier:  Thermo Scientific Chemicals 112950500

 View more versions of this product

Catalog No. AC112950500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-n-butylamine
-62.0°C
159.0°C
Authentic
Glass bottle
1.4160 to 1.4180
MFCD00009429
04, 157
0.76
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
JQVDAXLFBXTEQA-UHFFFAOYSA-N
N-butylbutan-1-amine
8148
129.24
Liquid
111-92-2
0.7600g/mL
39°C
98.5% min. (GC)
C8H19N
[CH3(CH2)3]2NH
50 mL
01,209
15, 3037
Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
CCCCNCCCC
129.24
0.9 mPa.s (20°C)
99%

Chemical Identifiers

111-92-2
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
CCCCNCCCC
MFCD00009429
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
N-butylbutan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre

GHS Signal Word: Danger

EINECSNumber : 203-921-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only