Learn More
Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals
As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions
$73.78 - $573.42
Chemical Identifiers
CAS | 1067-33-0 |
---|---|
Molecular Formula | C12H26O4Sn |
Molecular Weight (g/mol) | 353.05 |
MDL Number | MFCD00008697 |
InChI Key | LDZDQDDROJXUBO-UHFFFAOYSA-N |
Synonym | Diacetoxydi-n-butyltin |
SMILES | CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA1859518
|
Thermo Scientific Chemicals
01859518 |
50 g |
Each for $73.78
|
|
||||||
AA1859530
|
Thermo Scientific Chemicals
01859530 |
250 g |
Each for $224.62
|
|
||||||
AA18595A1
|
Thermo Scientific Chemicals
018595A1 |
1 kg |
Each for $573.42
|
|
||||||
Description
As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1067-33-0 | |
353.05 | |
LDZDQDDROJXUBO-UHFFFAOYSA-N | |
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC |
C12H26O4Sn | |
MFCD00008697 | |
Diacetoxydi-n-butyltin |
Specifications
1067-33-0 | |
Colorless to Yellow | |
142°C to 145°C (10 mmHg) | |
Acidic | |
1.47 | |
MFCD00008697 | |
UN2788 | |
Soluble in water and most organic solvents; Miscible with most organic solvents. | |
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC | |
351.01 | |
Liquid |
12°C | |
1.32 g/mL | |
143°C (290°F) | |
C12H26O4Sn | |
[OOCCH3]2Sn[(CH2)3CH3]2 | |
50 g | |
Diacetoxydi-n-butyltin | |
LDZDQDDROJXUBO-UHFFFAOYSA-N | |
353.05 | |
95% | |
Di-n-butyltin diacetate |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber : 213-928-8
RTECSNumber : WH6880000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only