missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals

$62.23 - $192.71

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1449122
View Documents
Thermo Scientific Chemicals
L1449122
100 g
Each for $62.23
 
AAL1449136
View Documents
Thermo Scientific Chemicals
L1449136
500 g
Each for $192.71
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

818-08-6
248.941
JGFBRKRYDCGYKD-UHFFFAOYSA-N
61221
CCCC[Sn](=O)CCCC
C8H18OSn
MFCD00001992
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
dibutyl(oxo)tin

Specifications

818-08-6
1.58
C8H18OSn
MFCD00001992
UN3146
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
CCCC[Sn](=O)CCCC
248.941
248.94
Di-n-butyltin oxide
>300°C
Odorless
[CH3(CH2)3]2SnO
100 g
4126243
JGFBRKRYDCGYKD-UHFFFAOYSA-N
dibutyl(oxo)tin
61221
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.

P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P307+P311-P310-P330-P333+P313-P362-P501c

H301-H315-H317-H318-H341-H360-H370-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 212-449-1

RTECSNumber : WH7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only