Learn More
Di-n-hexyl ether, 98%, Thermo Scientific Chemicals
$64.03 - $231.71
Chemical Identifiers
CAS | 112-58-3 |
---|---|
Molecular Formula | C12H26O |
Molecular Weight (g/mol) | 186.34 |
MDL Number | MFCD00009525 |
InChI Key | BPIUIOXAFBGMNB-UHFFFAOYSA-N |
Synonym | dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane |
PubChem CID | 8198 |
SMILES | CCCCCCOCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0707609
|
Thermo Scientific Chemicals
L0707609 |
10 g |
Each for $64.03
|
|
|||||
AAL0707618
|
Thermo Scientific Chemicals
L0707618 |
50 g |
Each for $231.71
|
|
|||||
Description
Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
112-58-3 | |
186.34 | |
BPIUIOXAFBGMNB-UHFFFAOYSA-N | |
8198 |
C12H26O | |
MFCD00009525 | |
dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane | |
CCCCCCOCCCCCC |
Specifications
112-58-3 | |
0.793 | |
78°C (172°F) | |
C12H26O | |
CH3(CH2)5O(CH2)5CH3 | |
10 g | |
dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane | |
BPIUIOXAFBGMNB-UHFFFAOYSA-N | |
1-hexoxyhexane | |
8198 | |
98% |
-43°C | |
228°C to 230°C | |
Mild | |
1.4204 | |
MFCD00009525 | |
1738177 | |
Fully miscible in water. | |
CCCCCCOCCCCCC | |
186.34 | |
186.34 | |
Di-n-hexyl ether |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 203-987-8
RTECSNumber : KN8600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only