Learn More
Di-n-octyl sulfide, 97%, Thermo Scientific Chemicals
$80.52 - $748.74
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1795514
|
Thermo Scientific Chemicals
A1795514 |
25 g |
Each for $80.52
|
|
|||||
AAA1795522
|
Thermo Scientific Chemicals
A1795522 |
100 g |
Each for $242.88
|
|
|||||
AAA1795536
|
Thermo Scientific Chemicals
A1795536 |
500 g |
Each for $748.74
|
|
|||||
Description
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a pharmaceutical intermediate.
Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)
Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
2690-08-6 | |
MFCD00009566 |
Specifications
2690-08-6 | |
0.844 | |
160°C (320°F) | |
C16H34S | |
MFCD00009566 | |
1701624 | |
LOXRGHGHQYWXJK-UHFFFAOYSA-N | |
1-octylsulfanyloctane | |
75901 | |
97% |
0°C to 1°C | |
179°C to 180°C (10 mmHg) | |
Stench | |
1.4624 | |
25 g | |
dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane | |
CCCCCCCCSCCCCCCCC | |
258.51 | |
258.51 | |
Di-n-octyl sulfide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-259-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only