missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octylamine, 96%

CAS: 1120-48-5 | C16H35N | 241.463 g/mol

$64.26 - $174.41

Chemical Identifiers

CAS 1120-48-5
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009557
InChI Key LAWOZCWGWDVVSG-UHFFFAOYSA-N
Synonym dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
PubChem CID 3094
IUPAC Name N-octyloctan-1-amine
SMILES CCCCCCCCNCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1801714
View Documents
Thermo Scientific Chemicals
A1801714
25 g N/A
Only null left
 
AAA1801722
View Documents
Thermo Scientific Chemicals
A1801722
100 g N/A
Only null left
 
Description

Description

Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.

Solubility
Immiscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications

Chemical Identifiers

1120-48-5
241.463
LAWOZCWGWDVVSG-UHFFFAOYSA-N
3094
CCCCCCCCNCCCCCCCC
C16H35N
MFCD00009557
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
N-octyloctan-1-amine

Specifications

1120-48-5
0.799
142°C (287°F)
C16H35N
[CH3(CH2)7]2NH
25 g
1748376
Immiscible in water.
CCCCCCCCNCCCCCCCC
241.463
241.46
Di-n-octylamine
13°C to 15°C
297°C to 298°C
Amine-like
1.4432
MFCD00009557
UN2735
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine
LAWOZCWGWDVVSG-UHFFFAOYSA-N
N-octyloctan-1-amine
3094
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 214-311-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only