missing translation for 'onlineSavingsMsg'
Learn More

Di-n-pentyl phthalate, 98%

CAS: 131-18-0 | C18H26O4 | 306.402 g/mol

$217.14 - $2157.21

Chemical Identifiers

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.402
MDL Number MFCD00041934
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
Synonym dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
PubChem CID 8561
ChEBI CHEBI:34680
IUPAC Name dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2291006
View Documents
Thermo Scientific Chemicals
B2291006
5 g
Each for $217.14
Only null left
 
AAB2291014
View Documents
Thermo Scientific Chemicals
B2291014
25 g
Each for $707.89
Only null left
 
AAB2291022
undefined
Thermo Scientific Chemicals
B2291022
100 g
Each for $2,157.21
Only null left
 
Description

Description

Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-pentyl phthalate is a plasticizer for PVC membrane electrodes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

131-18-0
306.402
IPKKHRVROFYTEK-UHFFFAOYSA-N
8561
dipentyl benzene-1,2-dicarboxylate
C18H26O4
MFCD00041934
dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
CHEBI:34680
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Specifications

131-18-0
342°C
Odorless
MFCD00041934
UN3082
Not miscible or difficult to mix in water.
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
306.402
CHEBI:34680
98%
1.025
118°C (244°F)
C18H26O4
5 g
dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
IPKKHRVROFYTEK-UHFFFAOYSA-N
dipentyl benzene-1,2-dicarboxylate
8561
306.41
Di-n-pentyl phthalate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P280-P308+P313-P501c

H360FD

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 205-017-9

RTECSNumber : TI1930000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only