Learn More
Di-n-propylamine, 99%, Thermo Scientific Chemicals
$61.25 - $61.25
Chemical Identifiers
CAS | 142-84-7 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.19 |
MDL Number | MFCD00009362 |
InChI Key | WEHWNAOGRSTTBQ-UHFFFAOYSA-N |
Synonym | dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805 |
PubChem CID | 8902 |
IUPAC Name | N-propylpropan-1-amine |
SMILES | CCCNCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL15808AP
|
Thermo Scientific Chemicals
L15808AP |
500 mL |
Each for $61.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-84-7 | |
101.19 | |
WEHWNAOGRSTTBQ-UHFFFAOYSA-N | |
8902 | |
CCCNCCC |
C6H15N | |
MFCD00009362 | |
dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805 | |
N-propylpropan-1-amine |
Specifications
142-84-7 | |
0.738 | |
7°C (45°F) | |
C6H15N | |
MFCD00009362 | |
UN2383 | |
14,3343 | |
WEHWNAOGRSTTBQ-UHFFFAOYSA-N | |
N-propylpropan-1-amine | |
8902 | |
99% |
-63°C | |
109°C to 110°C | |
Ammonia-like | |
1.405 | |
500 mL | |
505974 | |
dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805 | |
CCCNCCC | |
101.19 | |
101.19 | |
Di-n-propylamine |
Safety and Handling
GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311+H331-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIPROPYLAMINE
EINECSNumber : 205-565-9
RTECSNumber : JL9200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only