missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di(N-succinimidyl) Carbonate 98.0+%, TCI America™
Supplier: TCI America D166225G
Specifications
Di(N-succinimidyl) Carbonate | |
215°C | |
MFCD00009767 | |
n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700 | |
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O | |
256.17 | |
256.17 | |
Crystalline Powder |
74124-79-1 | |
C9H8N2O7 | |
25 g | |
PFYXSUNOLOJMDX-UHFFFAOYSA-N | |
bis(2,5-dioxopyrrolidin-1-yl) carbonate | |
676246 | |
≥98.0% (T) |
Chemical Identifiers
74124-79-1 | |
256.17 | |
PFYXSUNOLOJMDX-UHFFFAOYSA-N | |
676246 | |
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O |
C9H8N2O7 | |
MFCD00009767 | |
n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700 | |
bis(2,5-dioxopyrrolidin-1-yl) carbonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator