missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™, Thermo Scientific™

Specifications

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426801000
View Documents
Thermo Scientific Chemicals
426801000
100mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC426808000
View Documents
Thermo Scientific Chemicals
426808000
800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

24424-99-5
218.25
DYHSDKLCOJIUFX-UHFFFAOYSA-N
90495
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
C10H18O5
MFCD00008805
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CHEBI:48500
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only