Learn More
Di-tert-butyl malonate, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals
$122.18 - $1769.56
Chemical Identifiers
CAS | 541-16-2 |
---|---|
Molecular Formula | C11H20O4 |
Molecular Weight (g/mol) | 216.277 |
MDL Number | MFCD00008810 |
InChI Key | CLPHAYNBNTVRDI-UHFFFAOYSA-N |
Synonym | di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate |
PubChem CID | 68324 |
IUPAC Name | ditert-butyl propanedioate |
SMILES | CC(C)(C)OC(=O)CC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1277409
|
Thermo Scientific Chemicals
A1277409 |
10 g |
Each for $122.18
|
|
|||||
AAA1277418
|
Thermo Scientific Chemicals
A1277418 |
50 g |
Each for $427.33
|
|
|||||
AAA1277430
|
Thermo Scientific Chemicals
A1277430 |
250 g |
Each for $1,769.56
|
|
|||||
Description
Di-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-tert-butyl malonate, is used as a pharmaceutical intermediate. it can be used to produce Diazomalonsaeure-di-tert-butylester at the ambient temperature. It will need reagents cesium carbonate, p-toluenesulfonyl azide and solvent tetrahydrofuran with the reaction time of 1 hour. T
Solubility
It is hardly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Chemical Identifiers
541-16-2 | |
216.277 | |
CLPHAYNBNTVRDI-UHFFFAOYSA-N | |
68324 | |
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C |
C11H20O4 | |
MFCD00008810 | |
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
ditert-butyl propanedioate |
Specifications
541-16-2 | |
0.965 | |
88°C (190°F) | |
1.419 | |
10 g | |
14,3034 | |
It is hardly soluble in water. | |
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C | |
216.277 | |
216.28 | |
Di-tert-butyl malonate, stabilized with potassium carbonate |
-6°C | |
65°C to 67°C (1 mmHg) | |
C11H20O4 | |
MFCD00008810 | |
1781766 | |
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
CLPHAYNBNTVRDI-UHFFFAOYSA-N | |
ditert-butyl propanedioate | |
68324 | |
≥98% |
Safety and Handling
GHS H Statement
H317-H227
May cause an allergic skin reaction.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 208-769-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only