missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-tert-butyl Peroxide 98.0+%, TCI America™

Supplier:  TCI America D3411100ML

Catalog No. D3411100ML


Explore available promotions

Description

Description

Specifications

Specifications

Di-tert-butyl Peroxide
-40°C
111°C
MFCD00008803
3107
LSXWFXONGKSEMY-UHFFFAOYSA-N
2-(tert-butylperoxy)-2-methylpropane
8033
≥98.0% (GC)
110-05-4
Yellow
C8H18O2
100 mL
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
CC(C)(C)OOC(C)(C)C
146.23
146.23
Liquid

Chemical Identifiers

110-05-4
146.23
LSXWFXONGKSEMY-UHFFFAOYSA-N
8033
CC(C)(C)OOC(C)(C)C
C8H18O2
MFCD00008803
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
2-(tert-butylperoxy)-2-methylpropane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0367

RTECSNumber : ER2450000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions