missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Diacetone-D-glucose, 98+%
Supplier: Thermo Scientific Chemicals 112010250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diacetone-D-glucose | |
582-52-5 | |
White to Yellow | |
Authentic | |
Plastic bottle | |
MFCD00005544 | |
31,155 | |
− 11.00 (20.00°C c=5,EtoH) | |
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol | |
KEJGAYKWRDILTF-MQIGXGKASA-N | |
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol | |
40469256 | |
98+% |
98+% | |
107.0°C to 110.0°C | |
0.5% max. | |
98% min. (GC) | |
C12H20O6 | |
25 g | |
15,2968 | |
− 11.00 | |
Solubility in water: soluble. Other solubilities: freely soluble in chloroform,alcohol,acetone,and warm ether | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C | |
260.28 | |
260.28 | |
Powder |
Chemical Identifiers
582-52-5 | |
260.28 | |
KEJGAYKWRDILTF-MQIGXGKASA-N | |
40469256 | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C |
C12H20O6 | |
MFCD00005544 | |
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol | |
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol |
Safety and Handling
EINECSNumber : 209-486-0
RTECSNumber : LZ4958000
TSCA : TSCA
RUO – Research Use Only