missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diacetyl 98.0+%, TCI America™

Supplier:  TCI America B0682500ML

 View more versions of this product

Catalog No. B0682500ML


Explore available promotions

Description

Description

Specifications

Specifications

Diacetyl
-4°C
3.2 (50g/L Soln.H2O)
C4H6O2
500 mL
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
CC(=O)C(=O)C
86.09
CHEBI:16583
≥98.0% (GC)
431-03-8
Yellow
88°C
MFCD00008756
2346
QSJXEFYPDANLFS-UHFFFAOYSA-N
butane-2,3-dione
650
86.09
Liquid

Chemical Identifiers

431-03-8
86.09
QSJXEFYPDANLFS-UHFFFAOYSA-N
650
butane-2,3-dione
C4H6O2
MFCD00008756
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
CHEBI:16583
CC(=O)C(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0563

RTECSNumber : EK2625000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions